Fluorobenzene, 99%
Fluorobenzene, 99%
Fluorobenzene, 99%
Thermo Scientific Chemicals

Fluorobenzene, 99%

CAS: 462-06-6 | C6H5F | 96.10 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 119340025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS462-06-6
IUPAC Namefluorobenzene
Molecular FormulaC6H5F
InChI KeyPYLWMHQQBFSUBP-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1
さらに表示
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4643 to 1.4663 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.1 % (K.F.)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only