2-Furoic acid, 98%
2-Furoic acid, 98%
2-Furoic acid, 98%
Thermo Scientific Chemicals

2-Furoic acid, 98%

CAS: 88-14-2 | C5H4O3 | 112.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 119820050
Price (USD)
21.65
Special offer
Online exclusive
Ends: 28-Dec-2026
25.10
Save 3.45
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
21.65
Special offer
Online exclusive
Ends: 28-Dec-2026
25.10
Save 3.45
Each
Chemical Identifiers
CAS88-14-2
IUPAC Namefuran-2-carboxylic acid
Molecular FormulaC5H4O3
InChI KeySMNDYUVBFMFKNZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
Melting point129°C to 133°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only