D-Glucosamine hydrochloride, 98+%
D-Glucosamine hydrochloride, 98+%
D-Glucosamine hydrochloride, 98+%
Thermo Scientific Chemicals

D-Glucosamine hydrochloride, 98+%

数量:
10 g
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 119901000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS66-84-2
IUPAC Namehydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
Molecular FormulaC6H14ClNO5
InChI KeyCBOJBBMQJBVCMW-UHFFFAOYNA-N
SMILES[H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO
さらに表示
Appearance (Form)Crystalline powder
Titration with HClO498.0 to 102.0 % (On dry substance)
Arsenic (As)=<3 ppm
Specific optical rotation(after 3 h)
Specific optical rotation+70.0° to +73.0° (20°C, 589 nm) (c=2.5, H2O)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only