This product ships via ground from US northeast.
L(+)-Glutamic acid monosodium salt monohydrate, 99%
L(+)-Glutamic acid monosodium salt monohydrate, 99%
L(+)-Glutamic acid monosodium salt monohydrate, 99%
Thermo Scientific Chemicals

L(+)-Glutamic acid monosodium salt monohydrate, 99%

CAS: 6106-04-03 | C5H8NNaO4·H2O | 187.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 119940025
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS6106-04-3
IUPAC Namesodium (4S)-4-amino-4-carboxybutanoate hydrate
Molecular FormulaC5H10NNaO5
InChI KeyGJBHGUUFMNITCI-QTNFYWBSSA-M
SMILESO.[Na+].N[C@@H](CCC([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.5 % (100°C, 5 hrs)
Specific optical rotation+24.5° to +25.5° (20°C, 589 nm) (c=10, 2 N HCl)
Chloride (Cl)=<2000 ppm
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only