Heptaldehyde, 95%, stabilized
Heptaldehyde, 95%, stabilized
Heptaldehyde, 95%, stabilized
Thermo Scientific Chemicals

Heptaldehyde, 95%, stabilized

CAS: 111-71-7 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 120322500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range94% min. (GC)
Beilstein01, II, 750
CAS111-71-7
ChEBICHEBI:34787
Chemical Name or MaterialHeptaldehyde
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only