DMEM, powder, high glucose
Gibco™

DMEM, powder, high glucose

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySource
1210004610 x 1 LPrimary site of manufacture: Grand Island
1210006110 LPrimary site of manufacture: Grand Island
Catalog number 12100046
Price (USD)
70.65
Online exclusive
73.75
Save 3.10
Each
Quantity:
10 x 1 L
Source:
Primary site of manufacture: Grand Island
Customize this product
Price (USD)
70.65
Online exclusive
73.75
Save 3.10
Each

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: High Glucose, L-glutamine, Phenol Red

Without: Sodium Pyruvate, HEPES, Sodium Bicarbonate

The complete formulation is available.

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.

DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

A note about shelf-life: The shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHeLa, 293, Cos-7, and PC-12
Cell TypePrimary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeDMEM (Dulbecco's Modified Eagle Medium)
Quantity10 x 1 L
Shelf Life36 Months From Date of Manufacture
Shipping ConditionRoom Temperature
SourcePrimary site of manufacture: Grand Island
ClassificationAnimal Origin-free
FormPowder
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesHigh Glucose, Glutamine, Phenol Red
Without AdditivesNo HEPES, No Sodium Pyruvate, No Sodium Bicarbonate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Frequently asked questions (FAQs)

What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?

Manganese is not present in the formulation of our catalog DMEM media products.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the shelf life of my powdered media once I reconstitute it?

Expiration date of most Gibco reconstituted dry format media (AGT or DPM) has not been established; end users should assess performance and stability of this reconstituted media in a system that is relevant to their process. A dry format product (either DPM or AGT) will age upon storage and while we may not be able to detect which component(s) degrade since we can't measure every component, that doesn't mean that the reconstituted liquid will have the same stability as a fresh liquid made by direct weigh methods.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (5)

Citations & References
Abstract
Galectin-3 Translocates to the Perinuclear Membranes and Inhibits Cytochrome c Release from the Mitochondria. A ROLE FOR SYNEXIN IN GALECTIN-3 TRANSLOCATION.
Authors: Yu Fei; Finley Russell L Jr; Raz Avraham; Kim Hyeong-Reh Choi;
Journal:J Biol Chem
PubMed ID:11839755
'Galectin-3 is a multifunctional oncogenic protein found in the nucleus and cytoplasm and also the extracellular milieu. Although recent studies demonstrated an anti-apoptotic activity of galectin-3, neither the functional site nor the mechanism of how galectin-3 regulates apoptosis is known. In this study, we examined the subcellular localization of galectin-3 ... More
Functions of Transforming Growth Factor-beta Family Type I Receptors and Smad Proteins in the Hypertrophic Maturation and Osteoblastic Differentiation of Chondrocytes.
Authors: Valcourt Ulrich; Gouttenoire Jerome; Moustakas Aristidis; Herbage Daniel; Mallein-Gerin Frederic;
Journal:J Biol Chem
PubMed ID:12082094
'We investigated the effects of bone morphogenetic protein (BMP)-2, a member of the transforming growth factor-beta superfamily, on the regulation of the chondrocyte phenotype, and we identified signaling molecules involved in this regulation. BMP-2 triggers three concomitant responses in mouse primary chondrocytes and chondrocytic MC615 cells. First, BMP-2 stimulates expression ... More
Vasohibin 2 is transcriptionally activated and promotes angiogenesis in hepatocellular carcinoma.
Authors:Xue X, Gao W, Sun B, Xu Y, Han B, Wang F, Zhang Y, Sun J, Wei J, Lu Z, Zhu Y, Sato Y, Sekido Y, Miao Y, Kondo Y
Journal:Oncogene
PubMed ID:22614011
'Hepatocellular carcinoma (HCC) typically relies on angiogenesis for its malignant behavior, including growth and metastasis. Vasohibin 2 (VASH2) was previously identified as an angiogenic factor, but its role in tumorigenesis is unknown. Using quantitative PCR and western blot analyses, we found that VASH2 is overexpressed in HCC cells and tissues. ... More
Lentiviral vector production, titration, and transduction of primary neurons.
Authors:Ding B, Kilpatrick DL
Journal:
PubMed ID:23681623
'Lentiviral vectors have become very useful tools for transgene delivery. Based on their ability to transduce both dividing and nondividing cells and to produce long-term transgene expression, lentiviruses have found numerous applications in the biomedical sciences, including developmental neuroscience. This protocol describes how to prepare lentiviral vectors by calcium phosphate ... More
Evidence that the transmembrane biogenesis of aquaporin 1 is cotranslational in intact mammalian cells.
Authors: Dohke Yoko; Turner R James;
Journal:J Biol Chem
PubMed ID:11884383
Most polytopic membrane proteins are believed to integrate into the membrane of the endoplasmic reticulum (ER) cotranslationally. However, recent studies with Xenopus oocytes and dog pancreatic microsomes have suggested that this is not the case for human aquaporin 1 (AQP1). These experiments indicate that membrane-spanning segments (MSSs) 2 and 4 ... More