4-Hydroxy-3-methoxybenzoic acid, 97+%
4-Hydroxy-3-methoxybenzoic acid, 97+%
4-Hydroxy-3-methoxybenzoic acid, 97+%
Thermo Scientific Chemicals

4-Hydroxy-3-methoxybenzoic acid, 97+%

CAS: 121-34-6 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 121350250
Price (USD)
43.65
Online exclusive
47.90
Save 4.25
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Online exclusive
47.90
Save 4.25
Each
Chemical Identifiers
CAS121-34-6
IUPAC Name4-hydroxy-3-methoxybenzoic acid
Molecular FormulaC8H8O4
InChI KeyWKOLLVMJNQIZCI-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.0 %
Melting point208°C to 214°C
Appearance (Color)White to yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only