3-(4-Hydroxyphenyl)propionic acid, 98%
3-(4-Hydroxyphenyl)propionic acid, 98%
3-(4-Hydroxyphenyl)propionic acid, 98%
Thermo Scientific Chemicals

3-(4-Hydroxyphenyl)propionic acid, 98%

CAS: 501-97-3 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 121750250
Price (EUR)
52,60
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
52,60
Each
Chemical Identifiers
CAS501-97-3
IUPAC Name3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC9H10O3
InChI KeyNMHMNPHRMNGLLB-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Form)Powder or granules
Melting point126°C to 130°C
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only