Thermo Scientific Chemicals

8-Hydroxyquinaldine, 98%, Thermo Scientific Chemicals

Catalog number: 121870250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

8-Hydroxyquinaldine, 98%, Thermo Scientific Chemicals

Catalog number: 121870250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
826-81-3
IUPAC Name
2-methylquinolin-8-ol
Molecular Formula
C10H9NO
InChI Key
NBYLBWHHTUWMER-UHFFFAOYSA-N
SMILES
CC1=CC=C2C=CC=C(O)C2=N1
Appearance (Color)
Beige to brown
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
Melting point
68°C to 73°C
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text