2-Imidazolidone hemihydrate, 99+%
2-Imidazolidone hemihydrate, 99+%
2-Imidazolidone hemihydrate, 99+%
Thermo Scientific Chemicals

2-Imidazolidone hemihydrate, 99+%

CAS: 121325-67-5 | C6H14N4O3 | 190.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Catalog number 122040025
Price (EUR)
163,00
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (EUR)
163,00
Each
Chemical Identifiers
CAS121325-67-5
IUPAC Namebis(imidazolidin-2-one) hydrate
Molecular FormulaC6H14N4O3
InChI KeyKPRJGGOOWATRNT-UHFFFAOYSA-N
SMILESO.O=C1NCCN1.O=C1NCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder or crystals
Infrared spectrumConforms
Water=<13 %
Appearance (Color)White to light yellow
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only