2-Iodobenzoic acid, 98%
2-Iodobenzoic acid, 98%
2-Iodobenzoic acid, 98%
Thermo Scientific Chemicals

2-Iodobenzoic acid, 98%

CAS: 88-67-5 | C7H5IO2 | 248.01 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 122341000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS88-67-5
IUPAC Name2-iodobenzoic acid
Molecular FormulaC7H5IO2
InChI KeyCJNZAXGUTKBIHP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1I
さらに表示
Appearance (Color)White to off-white to beige
Melting point160°C to 164°C
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only