POROS™ 20 HQ Strong Anion Exchange Resin
El producto real puede variar
Thermo Scientific™

POROS™ 20 HQ Strong Anion Exchange Resin

Thermo Scientific™ POROS™ Strong Anion Exchange Media for scale-up of separations developed with POROS 20 micron Self Pack™ media.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
12329071000 mL
1232906
también denominado 1-2329-06
50 mL
Número de catálogo 1232907
Precio (MXN)
-
Cantidad:
1000 mL

POROS Strong Anion Exchange Media for scale-up of separations developed with POROS 20 micron Self Pack media. POROS 50 HQ is based on a quaternized polyethyleneimine functional group yielding a high capacity, High Performance Chromatography™ resins designed for the separation and purification of biomolecules.

POROS High Performance Chromatography™ Media and Pre-Packed Columns

A high performance chromatography resins for analytical to process scale separations.

  • Higher productivity: High throughput and high dynamic capacity associated with High Performance Chromatography™
  • Chemical stability: Allows aggressive cleaning and sanitization
  • Enhanced biomolecule access: Provided via large pores, ranging between 500-10,000 Å
  • Polystyrenedivinylbenzene particles: Yield a robust, easily packable matrix
  • Develop better separations methods in a shorter time frame: The speed of High Performance Chromatography technology reduces weeks of experimentation to only a few hours of work, so you have plenty of time to explore all the variables of your separation

Reduce time-consuming sample prep

You can replace many sample preparation steps with high speed High Performance Chromatography technology. For instance, dialysis of large volumes of material can be replaced by high flow rate processing of the dilute sample. Elution in a small volume of new buffer accomplishes both buffer exchange and concentration.

Create novel assays for more efficient analysis

High Performance Chromatography technology is not limited to standard modes of separation. Both enzymes and affinity ligands can be immobilized on POROS media. By combining rapid on-column protein digestions with rapid on-column immunoassays and chromatographic separations, you can create entirely new assays with unlimited potential.

High capacity, high resolution, high speed

In contrast to conventional chromatography media, POROS High Performance Chromatography™ resins particles, are engineered to have two discreet classes of pores. Large 'throughpores' allow convection flow to occur through the particles themselves, quickly carrying sample molecules to short 'diffusive' pores inside. By reducing the distance over which diffusion needs to occur, the time required for sample molecules to interact with interior binding sites is also reduced. Diffusion is no longer limiting and flow rates can be dramatically increased - without compromising resolution or capacity. Separations can be achieved at 1000 to 5000 cm/hr compared to 50 to 360 cm/hr for conventional media.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Fuerza de enlaceStrong
Tamaño de poro500 to 10,000 Å
Cantidad1000 mL
Condiciones de envíoTemperatura ambiente
Fase estacionariaHQ
FormularioMaciza
Línea de productosPOROS
TipoBulk Resin
Unit SizeEach
Contenido y almacenamiento
Almacenar a temperatura ambiente.