Aluminum sulfate octadecahydrate, extra pure
Aluminum sulfate octadecahydrate, extra pure
Aluminum sulfate octadecahydrate, extra pure
Thermo Scientific Chemicals

Aluminum sulfate octadecahydrate, extra pure

CAS: 7784-31-8 | Al2O12S3·18H2O | 666.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 123680025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7784-31-8
IUPAC Namedialuminium(3+) nonahydrate trisulfate
Molecular FormulaAl2H18O21S3
InChI KeyKFCWRPFNNIEZRY-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<20 ppm
Insoluble matter=<0.01 %
Chloride (Cl)=<50 ppm
Iron (Fe)=<50 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only