DL-Mandelic acid, 99+%
DL-Mandelic acid, 99+%
DL-Mandelic acid, 99+%
Thermo Scientific Chemicals

DL-Mandelic acid, 99+%

CAS: 90-64-2 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic Drum
Catalog number 125310050
Price (USD)
-
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS90-64-2
IUPAC Name2-hydroxy-2-phenylacetic acid
Molecular FormulaC8H8O3
InChI KeyIWYDHOAUDWTVEP-UHFFFAOYNA-N
SMILESOC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration with NaOH>=99.0 %
Appearance (Form)Crystalline powder or crystals
Melting point117°C to 121°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only