3-Methylthiophene, 99+%
3-Methylthiophene, 99+%
3-Methylthiophene, 99+%
Thermo Scientific Chemicals

3-Methylthiophene, 99+%

CAS: 616-44-4 | C5H6S | 98.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 127860500
Price (USD)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS616-44-4
IUPAC Name3-methylthiophene
Molecular FormulaC5H6S
InChI KeyQENGPZGAWFQWCZ-UHFFFAOYSA-N
SMILESCC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5180 to 1.5210 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only