1-Naphthol, 99+%
1-Naphthol, 99+%
1-Naphthol, 99+%
Acros Organics

1-Naphthol, 99+%

1-Naphthol, 99+%, C10H8O, CAS Number-90-15-3, 1-hydroxynaphthalene, 1-naphthalenol, 1-naphthol, alpha-naphthol, basf ursol ern, fouramine ern, fourrine ern, furro er, naphthol, tertral ern, 100g, 06, 596, 278.0 deg.C to 280.0 deg.C, 100.0, 99.0, CHEBI:10319, Beige-Gray to Brown, 201-969-4, 125 deg.C | CAS: 90-15-3 | C10H8O | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 128195000
HSN 29071510
Price (INR)
14,018.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
14,018.00
Each
Chemical Identifiers
CAS90-15-3
IUPAC Namenaphthalen-1-ol
Molecular FormulaC10H8O
InChI KeyKJCVRFUGPWSIIH-UHFFFAOYSA-N
SMILESOC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to grey to brown
Melting point95°C to 97°C
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only