1-Naphthyl isothiocyanate, 98%
1-Naphthyl isothiocyanate, 98%
1-Naphthyl isothiocyanate, 98%
Thermo Scientific Chemicals

1-Naphthyl isothiocyanate, 98%

CAS: 551-06-4 | C11H7NS | 185.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number 128240100
Price (USD)
157.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
157.00
Each
Chemical Identifiers
CAS551-06-4
IUPAC Name1-isothiocyanatonaphthalene
Molecular FormulaC11H7NS
InChI KeyJBDOSUUXMYMWQH-UHFFFAOYSA-N
SMILESS=C=NC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point53°C to 57°C
Appearance (Color)Yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only