2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
Thermo Scientific Chemicals

2-Nitrophenylhydrazine, 97%, moistened with ca 30% water

CAS: 3034-19-3 | C6H7N3O2 | 153.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
12883010010 gGlass bottle
Catalog number 128830100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3034-19-3
IUPAC Name2-(2-nitrophenyl)hydrazinium chloride
Molecular FormulaC6H8ClN3O2
InChI KeyXCUBVSAYUSFHNN-UHFFFAOYSA-N
SMILES[Cl-].[NH3+]NC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown to dark red
HPLC>=96.0 %
Infrared spectrumConforms
Water30 to 35 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only