Octadecylamine, 90%, tech.
Octadecylamine, 90%, tech.
Octadecylamine, 90%, tech.
Thermo Scientific Chemicals

Octadecylamine, 90%, tech.

CAS: 124-30-1 | C18H39N | 269.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 129320010
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.60
Save 14.95
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.60
Save 14.95
Each
Chemical Identifiers
CAS124-30-1
IUPAC Nameoctadecan-1-amine
Molecular FormulaC18H39N
InChI KeyREYJJPSVUYRZGE-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to cream
GC>=88 %
Amine number204 to 211 mg KOH/g (Total)
Melting point50°C to 60°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only