L(+)-Ornithine hydrochloride, 99%
L(+)-Ornithine hydrochloride, 99%
L(+)-Ornithine hydrochloride, 99%
Thermo Scientific Chemicals

L(+)-Ornithine hydrochloride, 99%

CAS: 3184-13-2 | C5H12N2O2·HCl | 168.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Catalog number 129560250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3184-13-2
IUPAC Namehydrogen 2,5-diaminopentanoic acid chloride
Molecular FormulaC5H13ClN2O2
InChI KeyGGTYBZJRPHEQDG-UHFFFAOYNA-N
SMILES[H+].[Cl-].NCCCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric>=98.5 %
Specific optical rotation+21° to +24° (20°C, 589 nm) (c=5, 1 N HCl)
Solubility(5 % in 1 N HCl) Clear colorless
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only