Oxalic acid dihydrate, 99+%, extra pure
Oxalic acid dihydrate, 99+%, extra pure
Oxalic acid dihydrate, 99+%, extra pure
Thermo Scientific Chemicals

Oxalic acid dihydrate, 99+%, extra pure

CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 129600050
Price (USD)
365.00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
365.00
Each
Chemical Identifiers
CAS6153-56-6
IUPAC Nameoxalic acid dihydrate
Molecular FormulaC2H6O6
InChI KeyGEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILESO.O.OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point98°C to 102°C
Titration with KMnO4>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only