Palmitic acid, 98%
Palmitic acid, 98%
Palmitic acid, 98%
Thermo Scientific Chemicals

Palmitic acid, 98%

CAS: 57-10-3 | C16H32O2 | 256.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
25 kg
Packaging:
View all
Fibre drum
Catalog number 129700250
Price (USD)
-
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Chemical Identifiers
CAS57-10-3
IUPAC Namehexadecanoic acid
Molecular FormulaC16H32O2
InChI KeyIPCSVZSSVZVIGE-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Acid value215 to 233 mg KOH/g
Iodine value=<1 g I2/100 g
Appearance (Form)Powder or granules or flakes
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only