D(-)-Penicillamine, 99%, specified according to Ph.Eur. and USP
D(-)-Penicillamine, 99%, specified according to Ph.Eur. and USP
D(-)-Penicillamine, 99%, specified according to Ph.Eur. and USP
Thermo Scientific Chemicals

D(-)-Penicillamine, 99%, specified according to Ph.Eur. and USP

CAS: 52-67-5 | C5H11NO2S | 149.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 129740050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52-67-5
IUPAC Name(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
Molecular FormulaC5H11NO2S
InChI KeyVVNCNSJFMMFHPL-VKHMYHEASA-N
SMILESCC(C)(S)[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Identification CPasses test
Specific optical rotation-60.5° to -65.0° (20°C, 589 nm) (0.5g in 10 ml 1N NaOH)
Mercury (Hg)=<0.001 %
pH4.5 to 5.5 (1 g in 100 ml water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only