Product Image
Thermo Scientific Chemicals

Phenylacetic acid, 98.5%

CAS: 103-82-2 | C8H8O2 | 136.15 g/mol
製品番号(カタログ番号) 130280050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS103-82-2
Titration with NaOH98.0 to 102.0 %
Appearance (Form)Crystalline powder or flakes
Melting point75°C to 78°C
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only