L-Phenylalanine, 98.5+%
L-Phenylalanine, 98.5+%
L-Phenylalanine, 98.5+%
Thermo Scientific Chemicals

L-Phenylalanine, 98.5+%

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number 130310250
Price (USD)
43.00
Each
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
43.00
Each
Chemical Identifiers
CAS63-91-2
IUPAC Name2-amino-3-phenylpropanoic acid
Molecular FormulaC9H11NO2
InChI KeyCOLNVLDHVKWLRT-UHFFFAOYNA-N
SMILESNC(CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.3 % (105°C, 3 hrs)
Specific optical rotation-33.0° to -35.2° (20°C, 589 nm) (c=2, water)
pH5.4 to 6.0 (1 % in water)
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only