3-Picoline, 99%
3-Picoline, 99%
3-Picoline, 99%
Thermo Scientific Chemicals

3-Picoline, 99%

CAS: 108-99-6 | C6H7N | 93.13 g/mol
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 131130010
Price (JPY)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-99-6
IUPAC Name3-methylpyridine
Molecular FormulaC6H7N
InChI KeyITQTTZVARXURQS-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear yellow
Refractive index1.5050 to 1.5070 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only