Picolinic acid, 99%
Picolinic acid, 99%
Picolinic acid, 99%
Thermo Scientific Chemicals

Picolinic acid, 99%

CAS: 98-98-6 | C6H5NO2 | 123.11 g/mol
数量:
100 g
500 g
2.5 kg
製品番号(カタログ番号) 131175000
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS98-98-6
IUPAC Namepyridine-2-carboxylic acid
Molecular FormulaC6H5NO2
InChI KeySIOXPEMLGUPBBT-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=N1
さらに表示
Titration with NaOH98.5 to 101.5 %
Appearance (Color)White to light yellow
Melting point136°C to 139°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only