Picrotoxin, 95%
Picrotoxin, 95%
Picrotoxin, 95%
Thermo Scientific Chemicals

Picrotoxin, 95%

CAS: 124-87-8 | C30H34O13 | 602.58 g/mol
数量:
1 g
5 g
25 g
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 131210250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS124-87-8
IUPAC Name(1R,3R,5S,8S,13R,14S)-1-hydroxy-14-(2-hydroxypropan-2-yl)-13-methyl-4,7,10-trioxapentacyclo[6.4.1.1⁹,¹².0³,⁵.0⁵,¹³]tetradecane-6,11-dione; (1R,5S,8S,13R,14R)-1-hydroxy-13-methyl-14-(prop-1-en-2-yl)-4,7,10-trioxapentacyclo[6.4.1.1⁹,¹².0³,⁵.0⁵,¹³]tetradecane-6,11-dione
Molecular FormulaC30H34O13
InChI KeyVJKUPQSHOVKBCO-ZTYBEOBUSA-N
SMILESCC(=C)[C@@H]1C2OC(=O)C1[C@]1(O)CC3O[C@]33C(=O)O[C@H]2[C@]13C.CC(C)(O)[C@@H]1C2OC(=O)C1[C@]1(O)C[C@H]3O[C@]33C(=O)O[C@H]2[C@]13C
さらに表示
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point198°C to 204°C
HPLC>=94 % (Mixture of Picrotoxinin and Picrotin)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only