3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals
3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals
3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals

数量:
100 mL
500 mL
2.5 L
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 131251000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS75-97-8
IUPAC Name3,3-dimethylbutan-2-one
Molecular FormulaC6H12O
InChI KeyPJGSXYOJTGTZAV-UHFFFAOYSA-N
SMILESCC(=O)C(C)(C)C
さらに表示
Infrared spectrumConforms
Refractive index1.3950 to 1.3980 (20°C, 589 nm)
GC>=95 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only