Propyl 4-hydroxybenzoate, 99+%
Propyl 4-hydroxybenzoate, 99+%
Propyl 4-hydroxybenzoate, 99+%
Propyl 4-hydroxybenzoate, 99+%
Thermo Scientific Chemicals

Propyl 4-hydroxybenzoate, 99+%

CAS: 94-13-3 | C10H12O3 | 180.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 131595000
Price (EUR)
106,00
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
106,00
Each
Add to cart
Chemical Identifiers
CAS94-13-3
IUPAC Namepropyl 4-hydroxybenzoate
Molecular FormulaC10H12O3
InChI KeyQELSKZZBTMNZEB-UHFFFAOYSA-N
SMILESCCCOC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance of solution(10% in ethanol 96%) clear colorless to
Appearance of solutionalmost colorless
Melting point95°C to 99°C
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only