Resorcinol, 98%
Resorcinol, 98%
Resorcinol, 98%
Thermo Scientific Chemicals

Resorcinol, 98%

CAS: 108-46-3 | C6H6O2 | 110.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 132292500
Price (USD)
72.70
Each
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
72.70
Each
Add to cart
Chemical Identifiers
CAS108-46-3
IUPAC Namebenzene-1,3-diol
Molecular FormulaC6H6O2
InChI KeyGHMLBKRAJCXXBS-UHFFFAOYSA-N
SMILESOC1=CC(O)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or granules or flakes
Melting point108°C to 112°C
Appearance (Color)White to yellow to beige to pink
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only