Rutin, 97+%
Rutin, 97+%
Rutin, 97+%
Thermo Scientific Chemicals

Rutin, 97+%

CAS: 207671-50-9 | C27H30O16 | 610.52 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 132395000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS207671-50-9
IUPAC Name2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-4H-chromen-4-one
Molecular FormulaC27H30O16
InChI KeyIKGXIBQEEMLURG-NVPNHPEKSA-N
SMILESC[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O
さらに表示
Appearance (Form)Powder
Appearance (Color)Yellow to yellow-green
Titration excess NaOH97.0 to 103.0 % (On dry basis)
Ash=<0.5 %
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only