Search
| Número do catálogo | Includes |
|---|---|
| 1324750 | 2-Port Laser Ablation Connection |
| BRE0017212 | 1-Port Laser Ablation Connection |
Expand your Analysis Capabilities with Advanced Accessories
Connect any laser ablation system to a Thermo Scientific™ iCAP™ Q/Qnova/MX Series ICP-MS system using our laser ablation connection kits.
Direct communication for Teledyne Photon Machines (including Teledyne CETAC Technologies) and Elemental Scientific Lasers (including NWR and ESI) laser ablation systems comes standard through dedicated plugins within the Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS) Software for truly routine laser ablation.
Full software support of quantification routines in laser ablation analysis is included in Qtegra ISDS Software.