Succinyl Chloride, ca. 95%
Succinyl Chloride, ca. 95%
Succinyl Chloride, ca. 95%
Thermo Scientific Chemicals

Succinyl Chloride, ca. 95%

CAS: 543-20-4 | C4H4Cl2O2 | 154.98 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 132840250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS543-20-4
IUPAC Namebutanedioyl dichloride
Molecular FormulaC4H4Cl2O2
InChI KeyIRXBNHGNHKNOJI-UHFFFAOYSA-N
SMILESClC(=O)CCC(Cl)=O
さらに表示
Appearance (Form)Liquid
Refractive index1.4670 to 1.4720 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Titration RCOCl>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only