4-Bromobutyric acid, 97%
4-Bromobutyric acid, 97%
4-Bromobutyric acid, 97%
Thermo Scientific Chemicals

4-Bromobutyric acid, 97%

CAS: 2623-87-2 | C4H7BrO2 | 167.00 g/mol
Have Questions?
Catalog NumberQuantityPackaging
133471000100 gGlass bottle
Catalog number 133471000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2623-87-2
IUPAC Name4-bromobutanoic acid
Molecular FormulaC4H7BrO2
InChI KeyGRHQDJDRGZFIPO-UHFFFAOYSA-N
SMILESOC(=O)CCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH96.0 to 104.0 %
Appearance (Form)Low melting crystals or fused solid
Infrared spectrumConforms
Melting point28°C to 33°C
Appearance (Color)White to yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only