4-Acetyl-4-phenylpiperidine hydrochloride, 99%, Thermo Scientific Chemicals
4-Acetyl-4-phenylpiperidine hydrochloride, 99%, Thermo Scientific Chemicals
4-Acetyl-4-phenylpiperidine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetyl-4-phenylpiperidine hydrochloride, 99%, Thermo Scientific Chemicals

CAS: 10315-03-4 | Molecular Formula: C13H17NO·ClH | Molecular Weight: 239.74 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1345800505 gGlass bottle
Catalog number 134580050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10315-03-4
IUPAC Name1-(4-phenylpiperidin-4-yl)ethan-1-one hydrochloridyl
Molecular FormulaC13H17ClNO
InChI KeyILIXCEOPJDISBX-UHFFFAOYSA-N
SMILES[Cl].CC(=O)C1(CCNCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
Appearance (Color)Light yellow to beige
Appearance (Form)Powder
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only