N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure
N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure
N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure

CAS: 110-18-9 | C6H16N2 | 116.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 138455000
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Chemical Identifiers
CAS110-18-9
IUPAC Name[2-(dimethylamino)ethyl]dimethylamine
Molecular FormulaC6H16N2
InChI KeyKWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILESCN(C)CCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly turbid colorless to yellow
Appearance (Form)Liquid
Refractive index1.4165 to 1.4195 (20°C, 589 nm)
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only