DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
Thermo Scientific Chemicals

DL-Thioctic Acid, 98+%

CAS: 1077-28-7 | C8H14O2S2 | 206.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 138720250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1077-28-7
IUPAC Name5-(1,2-dithiolan-3-yl)pentanoic acid
Molecular FormulaC8H14O2S2
InChI KeyAGBQKNBQESQNJD-UHFFFAOYNA-N
SMILESOC(=O)CCCCC1CCSS1
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrumComplies
Melting point60°C to 63°C
Appearance (Form)Crystalline powder or crystals
UV spectrophotometry98.0 to 102.0 % (on dry substance)
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only