2-Mercaptopropionic acid, 95%
2-Mercaptopropionic acid, 95%
2-Mercaptopropionic acid, 95%
Thermo Scientific Chemicals

2-Mercaptopropionic acid, 95%

CAS: 79-42-5 | C3H6O2S | 106.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 138791000
Price (EUR)
36,70
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
36,70
Each
Chemical Identifiers
CAS79-42-5
IUPAC Name2-sulfanylpropanoic acid
Molecular FormulaC3H6O2S
InChI KeyPMNLUUOXGOOLSP-UHFFFAOYNA-N
SMILESCC(S)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4804 to 1.4824 (20°C, 589 nm)
Appearance (Form)Liquid
Titration Iodimetric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only