2-Thiophenecarboxaldehyde, 98%
2-Thiophenecarboxaldehyde, 98%
2-Thiophenecarboxaldehyde, 98%
Thermo Scientific Chemicals

2-Thiophenecarboxaldehyde, 98%

CAS: 98-03-3 | C5H4OS | 112.15 g/mol
数量:
25 mL
100 mL
1 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 138830250
価格(JPY)
-
数量:
25 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS98-03-3
IUPAC Namethiophene-2-carbaldehyde
Molecular FormulaC5H4OS
InChI KeyCNUDBTRUORMMPA-UHFFFAOYSA-N
SMILESO=CC1=CC=CS1
さらに表示
Refractive index1.5890 to 1.5920 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only