m-Tolylacetic acid, 97%
m-Tolylacetic acid, 97%
m-Tolylacetic acid, 97%
Thermo Scientific Chemicals

m-Tolylacetic acid, 97%

CAS: 621-36-3 | C9H10O2 | 150.18 g/mol
数量:
5 g
25 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 139110050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS621-36-3
IUPAC Name2-(3-methylphenyl)acetic acid
Molecular FormulaC9H10O2
InChI KeyGJMPSRSMBJLKKB-UHFFFAOYSA-N
SMILESCC1=CC=CC(CC(O)=O)=C1
さらに表示
Infrared spectrumConforms
Appearance (Color)Colorless to off-white
Melting point61°C to 66°C
Appearance (Form)Glistening flakes and/or chunks
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only