Tributylamine, 99%
Tributylamine, 99%
Tributylamine, 99%
Thermo Scientific Chemicals

Tributylamine, 99%

CAS: 102-82-9 | C12H27N | 185.35 g/mol
Quantity:
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 139322500
Price (JPY)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS102-82-9
IUPAC Nametributylamine
Molecular FormulaC12H27N
InChI KeyIMFACGCPASFAPR-UHFFFAOYSA-N
SMILESCCCCN(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Color scale=<20 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4273 to 1.4293 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only