Trimethylacetonitrile, 98%
Trimethylacetonitrile, 98%
Trimethylacetonitrile, 98%
Thermo Scientific Chemicals

Trimethylacetonitrile, 98%

CAS: 630-18-2 | C5H9N | 83.13 g/mol
数量:
25 mL
100 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 140030250
価格(JPY)
-
数量:
25 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS630-18-2
IUPAC Name2,2-dimethylpropanenitrile
Molecular FormulaC5H9N
InChI KeyJAMNHZBIQDNHMM-UHFFFAOYSA-N
SMILESCC(C)(C)C#N
さらに表示
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3770 to 1.3790 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only