Triphenylmethyl mercaptan, 97%
Triphenylmethyl mercaptan, 97%
Triphenylmethyl mercaptan, 97%
Thermo Scientific Chemicals

Triphenylmethyl mercaptan, 97%

CAS: 3695-77-0 | C19H16S | 276.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 140411000
Price (USD)
222.65
Special offer
Online exclusive
Ends: 28-Dec-2026
262.00
Save 39.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
222.65
Special offer
Online exclusive
Ends: 28-Dec-2026
262.00
Save 39.35
Each
Chemical Identifiers
CAS3695-77-0
IUPAC Nametriphenylmethanethiol
Molecular FormulaC19H16S
InChI KeyJQZIKLPHXXBMCA-UHFFFAOYSA-N
SMILESSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point102°C to 109°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only