Urea, 99.5%, for analysis
Urea, 99.5%, for analysis
Urea, 99.5%, for analysis
Thermo Scientific Chemicals

Urea, 99.5%, for analysis

CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bag in resealable cardbox
Catalog number 140750250
Price (USD)
506.65
Special offer
Online exclusive
Ends: 28-Dec-2026
596.00
Save 89.35
Each
Quantity:
25 kg
Packaging:
Plastic bag in resealable cardbox
Request bulk or custom format
Price (USD)
506.65
Special offer
Online exclusive
Ends: 28-Dec-2026
596.00
Save 89.35
Each
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
Appearance (Color)White
Appearance (Form)Crystals
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only