(Carbethoxyethylidene)triphenylphosphorane, ca. 94%, balance TPPO
(Carbethoxyethylidene)triphenylphosphorane, ca. 94%, balance TPPO
(Carbethoxyethylidene)triphenylphosphorane, ca. 94%, balance TPPO
Thermo Scientific Chemicals

(Carbethoxyethylidene)triphenylphosphorane, ca. 94%, balance TPPO

CAS: 5717-37-3 | C23H23O2P | 362.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 146080250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5717-37-3
IUPAC Nameethyl 2-(triphenyl-λ⁵-phosphanylidene)propanoate
Molecular FormulaC23H23O2P
InChI KeyKZENFXVDPUMQOE-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLCca. 94 %
Infrared spectrumConforms
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only