Triphenylmethane, 99+%
Triphenylmethane, 99+%
Triphenylmethane, 99+%
Thermo Scientific Chemicals

Triphenylmethane, 99+%

CAS: 519-73-3 | C19H16 | 244.34 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 146381000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS519-73-3
Appearance (Color)Light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point90°C to 94°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only