Chlorobenzene, 99+%, pure
Chlorobenzene, 99+%, pure
Chlorobenzene, 99+%, pure
Thermo Scientific Chemicals

Chlorobenzene, 99+%, pure

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 146410010
Price (USD)
100.65
Online exclusive
112.00
Save 11.35
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
100.65
Online exclusive
112.00
Save 11.35
Each
Chemical Identifiers
CAS108-90-7
IUPAC Namechlorobenzene
Molecular FormulaC6H5Cl
InChI KeyMVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.0 %
Infrared spectrumConforms
Refractive index1.5230 to 1.5250 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only