Piperidine, 99%, extra pure
Thermo Scientific Chemicals

Piperidine, 99%, extra pure

CAS: 110-89-4 | C5H11N | 85.15 g/mol
数量:
500 mL
1 L
2.5 L
10 L
製品番号(カタログ番号) 147180010
価格(JPY)
-
数量:
1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-89-4
IUPAC Namepiperidine
Molecular FormulaC5H11N
InChI KeyNQRYJNQNLNOLGT-UHFFFAOYSA-N
SMILESC1CCNCC1
さらに表示
Infrared spectrumConforms
Appearance (Form)Clear liquid
Color scale=<50 APHA
Refractive index1.4510 to 1.4540 (20°C, 589 nm)
Water=<0.5 % (K.F.)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only