Phenolphthalein, 98.5%, pure, Indicator grade
Phenolphthalein, 98.5%, pure, Indicator grade
Phenolphthalein, 98.5%, pure, Indicator grade
Thermo Scientific Chemicals

Phenolphthalein, 98.5%, pure, Indicator grade

CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 147711000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS77-09-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC20H14O4
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
さらに表示
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Assay98.0 to 102.0 %
Loss on drying=<0.5 % (105°C)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only